missing translation for 'onlineSavingsMsg'
Learn More

2-Methylstyrene (stabilized with TBC) 97.0+%, TCI America™

Catalog No. M02595ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02595ML Supplier TCI America Supplier No. M02595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-15-4
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00014936
InChI Key NVZWEEGUWXZOKI-UHFFFAOYSA-N
Synonym 2-methylstyrene, 2-vinyltoluene, o-vinyltoluene, o-methylstyrene, styrene, o-methyl, 1-methyl-2-vinylbenzene, benzene, 1-ethenyl-2-methyl, ortho-methylstyrene, unii-556uo5cl44, 1-ethenyl-2-methyl-benzene
PubChem CID 11904
IUPAC Name 1-ethenyl-2-methylbenzene
SMILES CC1=CC=CC=C1C=C

Specifications

Melting Point -69°C
Color Yellow
Boiling Point 171°C
Quantity 5 mL
UN Number 2618
Formula Weight 118.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylstyrene (stabilized with TBC)
EINECSNumber (3)-0008
RTECSNumber WL5075900
TSCA No
Recommended Storage Refrigerator