accessibility menu, dialog, popup

UserName

CAS RN 611-15-4

IUPAC Name: 1-ethenyl-2-methylbenzene
Synonyms: O-methylstyrene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: NVZWEEGUWXZOKI-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)