missing translation for 'onlineSavingsMsg'
Learn More

2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. AAA1267606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1267606 5 g
AAA1267614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1267606 Supplier Thermo Scientific Chemicals Supplier No. A1267606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-15-4 | C9H10 | 118.179 g/mol

2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents and strong acids.

Chemical Identifiers

CAS 611-15-4
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00014936
InChI Key NVZWEEGUWXZOKI-UHFFFAOYSA-N
Synonym 2-methylstyrene, 2-vinyltoluene, o-vinyltoluene, o-methylstyrene, styrene, o-methyl, 1-methyl-2-vinylbenzene, benzene, 1-ethenyl-2-methyl, ortho-methylstyrene, unii-556uo5cl44, 1-ethenyl-2-methyl-benzene
PubChem CID 11904
IUPAC Name 1-ethenyl-2-methylbenzene
SMILES CC1=CC=CC=C1C=C

Specifications

Melting Point -69°C
Density 0.916
Boiling Point 170°C to 172°C
Flash Point 58°C (136°F)
Refractive Index 1.545
Quantity 5 g
UN Number UN2618
Beilstein 1901764
Solubility Information Slightly miscible with water.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 2-Methylstyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED
EINECSNumber 210-256-7
RTECSNumber WL5075900
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only