Learn More
2-Methylphenoxyacetic acid, 98%
CAS: 1878-49-5 | C9H10O3 | 166.176 g/mol
Supplier: Thermo Scientific Chemicals A1307618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methylphenoxyacetic acid | |
| 152°C to 156°C | |
| MFCD00014354 | |
| 1943465 | |
| QJVXBRUGKLCUMY-UHFFFAOYSA-N | |
| 2-(2-methylphenoxy)acetic acid | |
| 74651 | |
| 98% |
| 1878-49-5 | |
| C9H10O3 | |
| 50 g | |
| 2-methylphenoxy acetic acid, 2-2-methylphenoxy acetic acid, 2-methylphenoxyacetic acid, acetic acid, 2-methylphenoxy, o-tolyloxyacetic acid, o-tolyloxy acetic acid, 2-o-tolyloxy acetic acid, acetic acid, o-tolyloxy, acide o-methylphenoxyacetique, unii-pxo85n85g3 | |
| CC1=CC=CC=C1OCC(=O)O | |
| 166.176 | |
| 166.18 |
Chemical Identifiers
| 1878-49-5 | |
| 166.176 | |
| QJVXBRUGKLCUMY-UHFFFAOYSA-N | |
| 74651 | |
| CC1=CC=CC=C1OCC(=O)O |
| C9H10O3 | |
| MFCD00014354 | |
| 2-methylphenoxy acetic acid, 2-2-methylphenoxy acetic acid, 2-methylphenoxyacetic acid, acetic acid, 2-methylphenoxy, o-tolyloxyacetic acid, o-tolyloxy acetic acid, 2-o-tolyloxy acetic acid, acetic acid, o-tolyloxy, acide o-methylphenoxyacetique, unii-pxo85n85g3 | |
| 2-(2-methylphenoxy)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 217-517-4
RTECSNumber : AJ7573000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only