accessibility menu, dialog, popup

UserName

CAS RN 1878-49-5

IUPAC Name: 2-(2-methylphenoxy)acetic acid
Synonyms: O-methylphenoxyacetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: QJVXBRUGKLCUMY-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results