missing translation for 'onlineSavingsMsg'
Learn More

2-Methylphenoxyacetic acid, 98%

Catalog No. AAA1307630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1307630 250 g
AAA1307609 10 g
AAA1307618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1307630 Supplier Thermo Scientific Chemicals Supplier No. A1307630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1878-49-5 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-49-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014354
InChI Key QJVXBRUGKLCUMY-UHFFFAOYSA-N
Synonym 2-methylphenoxy acetic acid, 2-2-methylphenoxy acetic acid, 2-methylphenoxyacetic acid, acetic acid, 2-methylphenoxy, o-tolyloxyacetic acid, o-tolyloxy acetic acid, 2-o-tolyloxy acetic acid, acetic acid, o-tolyloxy, acide o-methylphenoxyacetique, unii-pxo85n85g3
PubChem CID 74651
IUPAC Name 2-(2-methylphenoxy)acetic acid
SMILES CC1=CC=CC=C1OCC(=O)O

Specifications

Melting Point 152°C to 156°C
Quantity 250 g
Beilstein 1943465
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Methylphenoxyacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-517-4
RTECSNumber AJ7573000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only