missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhydroquinone, 99%

Catalog No. AAB2124130
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2124130 250 g
AAB212410B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2124130 Supplier Thermo Scientific Chemicals Supplier No. B2124130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-71-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-71-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002345
InChI Key CNHDIAIOKMXOLK-UHFFFAOYSA-N
Synonym methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol
PubChem CID 7253
IUPAC Name 2-methylbenzene-1,4-diol
SMILES CC1=CC(O)=CC=C1O

Specifications

Melting Point 127°C to 130°C
Boiling Point 284°C to 286°C
Flash Point 172°C (341°F)
Quantity 250 g
Beilstein 2041489
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2-Methylhydroquinone
Hazard Category H302-H317-H318-H341-H351
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
EINECSNumber 202-443-7
RTECSNumber MX6700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only