missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 583-59-5 | C7H14O | 114.19 g/mol

Supplier:  Thermo Scientific Chemicals 166890010

 View more versions of this product

Catalog No. AC166890010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylcyclohexanol
98.5
-38.0°C
0.9300g/mL
58°C
98.5% min. (GC)
C7H14O
CH3C6H10OH
1 kg
08,303
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
NDVWOBYBJYUSMF-UHFFFAOYSA-N
2-methylcyclohexan-1-ol
11418
99%
583-59-5
100.0
Colorless
163.0°C to 166.0°C
Authentic
Glass bottle
1.4615 to 1.4635
MFCD00001443
06, 11
0.93
Solubility in water: slightly soluble
CC1CCCCC1O
114.19
114.19
Liquid

Chemical Identifiers

583-59-5
114.19
NDVWOBYBJYUSMF-UHFFFAOYSA-N
11418
CC1CCCCC1O
C7H14O
MFCD00001443
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
2-methylcyclohexan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.

GHS Signal Word: Warning

EINECSNumber : 209-512-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only