missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™
$170.66 - $295.81
Specifications
| CAS | 78-78-4 |
|---|---|
| Vapor Pressure | 11.17 psi (20°C) |
| Vapor Density | 2.6 (vs. air) |
| Density | 0.62 g/mL (at 25°C) |
| Physical Form | Neat |
Specifications
| 78-78-4 | |
| 2.6 (vs. air) | |
| Neat | |
| MFCD00009338 | |
| Isopentane | |
| ≥99.7% (GC) |
| 11.17 psi (20°C) | |
| 0.62 g/mL (at 25°C) | |
| CH3CH2CH(CH3)2 | |
| 30°C | |
| 72.15 | |
| Analytical Standard |