missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019821
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019821 5 mL
111018954 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019821 Supplier Merck Emd Millipore Supplier No. 590605ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 78-78-4
Vapor Pressure 11.17 psi (20°C)
Vapor Density 2.6 (vs. air)
Density 0.62 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH(CH3)2
MDL Number MFCD00009338
Boiling Point 30°C
Synonym Isopentane
Quantity 5 mL
Formula Weight 72.15
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less