missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl chloride, 98+%

CAS: 552-45-4 | C8H9Cl | 140.61 g/mol

Supplier:  Thermo Scientific Chemicals A1344822

 View more versions of this product

Catalog No. AAA1344822


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylbenzyl chloride
2°C
197°C to 199°C
Pungent
1.541
100 g
386016
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
CC1=CC=CC=C1CCl
140.61
140.61
552-45-4
1.081
73°C (163°F)
C8H9Cl
MFCD00000899
UN3265
14,10091
VQRBXYBBGHOGFT-UHFFFAOYSA-N
1-(chloromethyl)-2-methylbenzene
11093
≥98%

Chemical Identifiers

552-45-4
140.61
VQRBXYBBGHOGFT-UHFFFAOYSA-N
11093
CC1=CC=CC=C1CCl
C8H9Cl
MFCD00000899
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
1-(chloromethyl)-2-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-013-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only