missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbenzhydryl Chloride 98.0+%, TCI America™
$126.76 - $380.27
Chemical Identifiers
| CAS | 41870-52-4 |
|---|---|
| Molecular Formula | C14H13Cl |
| Molecular Weight (g/mol) | 216.71 |
| MDL Number | MFCD00972765 |
| InChI Key | VTZTUKIBJQCCMO-UHFFFAOYNA-N |
| Synonym | 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane |
| PubChem CID | 10798787 |
| IUPAC Name | 1-[chloro(phenyl)methyl]-2-methylbenzene |
| SMILES | CC1=CC=CC=C1C(Cl)C1=CC=CC=C1 |
Chemical Identifiers
| 41870-52-4 | |
| 216.71 | |
| VTZTUKIBJQCCMO-UHFFFAOYNA-N | |
| 10798787 | |
| CC1=CC=CC=C1C(Cl)C1=CC=CC=C1 |
| C14H13Cl | |
| MFCD00972765 | |
| 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane | |
| 1-[chloro(phenyl)methyl]-2-methylbenzene |
Specifications
| 41870-52-4 | |
| White-Yellow | |
| C14H13Cl | |
| 5 g | |
| 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane | |
| CC1=CC=CC=C1C(Cl)C1=CC=CC=C1 | |
| 216.71 | |
| 216.71 | |
| Crystalline Lumps |
| 40°C | |
| 136°C | |
| MFCD00972765 | |
| 3077 | |
| VTZTUKIBJQCCMO-UHFFFAOYNA-N | |
| 1-[chloro(phenyl)methyl]-2-methylbenzene | |
| 10798787 | |
| ≥98.0% (GC,T) | |
| 2-Methylbenzhydryl Chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator