accessibility menu, dialog, popup

UserName

CAS RN 41870-52-4

IUPAC Name: 1-[chloro(phenyl)methyl]-2-methylbenzene
Synonyms: 1-[chloro(phenyl)methyl]-2-methylbenzene
Molecular Weight (g/mol): 216.71
Molecular Formula: C14H13Cl
InChi Key: VTZTUKIBJQCCMO-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1C(Cl)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results