missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzhydryl Chloride 98.0+%, TCI America™

Catalog No. M22115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22115G 5 g
M221125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22115G Supplier TCI America Supplier No. M22115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41870-52-4
Molecular Formula C14H13Cl
Molecular Weight (g/mol) 216.71
MDL Number MFCD00972765
InChI Key VTZTUKIBJQCCMO-UHFFFAOYNA-N
Synonym 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane
PubChem CID 10798787
IUPAC Name 1-[chloro(phenyl)methyl]-2-methylbenzene
SMILES CC1=CC=CC=C1C(Cl)C1=CC=CC=C1

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
UN Number 3077
Formula Weight 216.71
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methylbenzhydryl Chloride
TSCA No
Recommended Storage Refrigerator