missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanisole, 99%

Catalog No. p-159374
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126281000 100 g Glass bottle
AC126285000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126281000 Supplier Thermo Scientific Chemicals Supplier No. 126281000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 578-58-5 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 578-58-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008373
InChI Key DTFKRVXLBCAIOZ-UHFFFAOYSA-N
Synonym 2-methylanisole, o-methylanisole, 2-methoxytoluene, o-cresol methyl ether, o-cresyl methyl ether, anisole, o-methyl, benzene, 1-methoxy-2-methyl, methyl o-tolyl ether, o-methoxytoluene, 2-methylmethoxybenzene
PubChem CID 33637
IUPAC Name 1-methoxy-2-methylbenzene
SMILES CC1=CC=CC=C1OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 170°C to 172°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.518
Linear Formula CH3C6H4OCH3
Quantity 100 g
Beilstein 06, 352
Specific Gravity 0.98
Solubility Information Solubility in water: immiscible
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylanisole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-426-3
TSCA TSCA

RUO – Research Use Only