accessibility menu, dialog, popup

UserName

CAS RN 578-58-5

IUPAC Name: 1-methoxy-2-methylbenzene
Synonyms: 2-methylanisole
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: DTFKRVXLBCAIOZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (6)
  • (2)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)