missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetanilide, 98+%

Catalog No. AAA1323522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323522 100 g
AAA1323514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1323522 Supplier Thermo Scientific Chemicals Supplier No. A1323522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-66-1 | C9H11NO | 149.193 g/mol

Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

Solubility
It is slightly soluble in acetone, chloroform, toluene, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under ordinary conditions.

Chemical Identifiers

CAS 120-66-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00014961
InChI Key BPEXTIMJLDWDTL-UHFFFAOYSA-N
Synonym o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide
PubChem CID 8443
IUPAC Name N-(2-methylphenyl)acetamide
SMILES CC1=CC=CC=C1NC(=O)C

Specifications

Melting Point 109°C to 112°C
Density 1.16
Boiling Point 296°C
Quantity 100 g
Beilstein 2207054
Merck Index 14,74
Solubility Information It is slightly soluble in acetone,chloroform,toluene,benzene.
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material 2'-Methylacetanilide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-414-4
RTECSNumber AN2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only