accessibility menu, dialog, popup

UserName

CH3ONHH3C

CAS RN 120-66-1

IUPAC Name: N-(2-methylphenyl)acetamide
Synonyms: acetamide, N-(2-methylphenyl)-
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: BPEXTIMJLDWDTL-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)