missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 312.38 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |