CAS RN 556812-44-3
CAS RN 556812-44-3
IUPAC Name:
2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms:
2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol):
312.38
Molecular Formula:
C11H15F3O3SSi
InChi Key:
ZCWWZFXNICWDIB-UHFFFAOYSA-N
SMILES:
CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C
1
–
1
of
1
results
1
2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate 96.0+%, TCI America™
CAS: 556812-44-3 Molecular Formula: C11H15F3O3SSi Molecular Weight (g/mol): 312.38 MDL Number: MFCD10566915 InChI Key: ZCWWZFXNICWDIB-UHFFFAOYSA-N Synonym: 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate PubChem CID: 12182646 IUPAC Name: 2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate SMILES: CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C