missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate 96.0+%, TCI America™

Catalog No. M18831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18831G 1 g
M18835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18831G Supplier TCI America Supplier No. M18831G
Only null left

Chemical Identifiers

CAS 556812-44-3
Molecular Formula C11H15F3O3SSi
Molecular Weight (g/mol) 312.38
MDL Number MFCD10566915
InChI Key ZCWWZFXNICWDIB-UHFFFAOYSA-N
Synonym 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate
PubChem CID 12182646
IUPAC Name 2-methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate
SMILES CC1=C(OS(=O)(=O)C(F)(F)F)C(=CC=C1)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 312.38
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-6-(trimethylsilyl)phenyl Trifluoromethanesulfonate