missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-ethylaniline 98.0+%, TCI America™

Catalog No. p-7132353
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M077225ML 25 mL
M0772500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M077225ML Supplier TCI America Supplier No. M077225ML

Chemical Identifiers

CAS 24549-06-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007752
InChI Key JJVKJJNCIILLRP-UHFFFAOYSA-N
Synonym 6-ethyl-o-toluidine, 2-methyl-6-ethylaniline, benzenamine, 2-ethyl-6-methyl, o-toluidine, 6-ethyl, 6-ethyl-2-toluidine, 2-methyl-6-ethyl aniline, aniline, 2-methyl-6-ethyl, 6-ethyl-2-methylaniline, 2-ethyl-6-methyl-aniline, 2-ethyl-6-methylbenzenamine
PubChem CID 32485
IUPAC Name 2-ethyl-6-methylaniline
SMILES CCC1=CC=CC(=C1N)C

Specifications

Melting Point -33°C
Boiling Point 231°C
Quantity 25 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methyl-6-ethylaniline
RTECSNumber BY5600000
TSCA Yes