accessibility menu, dialog, popup

UserName

CAS RN 24549-06-2

IUPAC Name: 2-ethyl-6-methylaniline
Synonyms: benzenamine, 2-ethyl-6-methyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: JJVKJJNCIILLRP-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(C)=C1N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results