missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. p-7133145
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23011G 1 g
M23015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23011G Supplier TCI America Supplier No. M23011G
Only null left

Chemical Identifiers

CAS 25449-96-1
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00134580
InChI Key BCLCKENDTZITFB-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzotrifluoride, 2-methyl-5-trifluoromethyl aniline, benzenamine, 2-methyl-5-trifluoromethyl, 2-methyl-5-trifluoromethyl phenylamine, pubchem4494, intermediates-zcf02681, acmc-209gk1, 5-trifluoromethyl-o-toluidine, 5-trifluoromethyl-2-methylaniline
PubChem CID 2735933
IUPAC Name 2-methyl-5-(trifluoromethyl)aniline
SMILES CC1=C(C=C(C=C1)C(F)(F)F)N

Specifications

Melting Point 40°C
Boiling Point 74°C
Quantity 1 g
Formula Weight 175.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methyl-5-(trifluoromethyl)aniline