Learn More
2-Methyl-5-(trifluoromethoxy)benzylamine, 96%, Thermo Scientific Chemicals
$423.67 - $423.67
Chemical Identifiers
| CAS | 1373920-87-6 |
|---|---|
| Molecular Formula | C9H10F3NO |
| Molecular Weight (g/mol) | 205.18 |
| MDL Number | MFCD22201056 |
| InChI Key | HUHITEUBBNSNER-UHFFFAOYSA-N |
| Synonym | 2-methyl-5-trifluoromethoxy benzylamine, 2-methyl-5-trifluoromethoxy phenyl methanamine, 1-2-methyl-5-trifluoromethoxy phenyl methanamine |
| PubChem CID | 86277657 |
| IUPAC Name | [2-methyl-5-(trifluoromethoxy)phenyl]methanamine |
| SMILES | CC1=C(CN)C=C(OC(F)(F)F)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3379103
|
Thermo Scientific Chemicals
H3379103 |
1 g |
Each for $423.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1373920-87-6 | |
| 205.18 | |
| HUHITEUBBNSNER-UHFFFAOYSA-N | |
| 86277657 | |
| CC1=C(CN)C=C(OC(F)(F)F)C=C1 |
| C9H10F3NO | |
| MFCD22201056 | |
| 2-methyl-5-trifluoromethoxy benzylamine, 2-methyl-5-trifluoromethoxy phenyl methanamine, 1-2-methyl-5-trifluoromethoxy phenyl methanamine | |
| [2-methyl-5-(trifluoromethoxy)phenyl]methanamine |
Specifications
| 1373920-87-6 | |
| MFCD22201056 | |
| UN3267 | |
| 2-methyl-5-trifluoromethoxy benzylamine, 2-methyl-5-trifluoromethoxy phenyl methanamine, 1-2-methyl-5-trifluoromethoxy phenyl methanamine | |
| CC1=C(CN)C=C(OC(F)(F)F)C=C1 | |
| 205.18 | |
| 205.18 | |
| 2-Methyl-5-(trifluoromethoxy)benzylamine |
| C9H10F3NO | |
| 1 g | |
| Air sensitive | |
| HUHITEUBBNSNER-UHFFFAOYSA-N | |
| [2-methyl-5-(trifluoromethoxy)phenyl]methanamine | |
| 86277657 | |
| 96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only