CAS RN 1373920-87-6
IUPAC Name:
1-[2-methyl-5-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[2-methyl-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
205.18
Molecular Formula:
C9H10F3NO
InChi Key:
HUHITEUBBNSNER-UHFFFAOYSA-N
SMILES:
CC1=C(CN)C=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Methyl-5-(trifluoromethoxy)benzylamine, 96%, Thermo Scientific Chemicals
2-Methyl-5-(trifluoromethoxy)benzylamine, 96%, Thermo Scientific Chemicals
CAS: 1373920-87-6 Molecular Formula: C9H10F3NO Molecular Weight (g/mol): 205.18 MDL Number: MFCD22201056 InChI Key: HUHITEUBBNSNER-UHFFFAOYSA-N Synonym: 2-methyl-5-trifluoromethoxy benzylamine,2-methyl-5-trifluoromethoxy phenyl methanamine,1-2-methyl-5-trifluoromethoxy phenyl methanamine PubChem CID: 86277657 IUPAC Name: [2-methyl-5-(trifluoromethoxy)phenyl]methanamine SMILES: CC1=C(CN)C=C(OC(F)(F)F)C=C1