missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)benzylamine, 96%, Thermo Scientific Chemicals

Catalog No. AAH3379103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3379103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3379103 Supplier Thermo Scientific Chemicals Supplier No. H3379103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1373920-87-6 | Molecular Formula: C9H10F3NO | Molecular Weight: 205.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1373920-87-6
Molecular Formula C9H10F3NO
Molecular Weight (g/mol) 205.18
MDL Number MFCD22201056
InChI Key HUHITEUBBNSNER-UHFFFAOYSA-N
Synonym 2-methyl-5-trifluoromethoxy benzylamine, 2-methyl-5-trifluoromethoxy phenyl methanamine, 1-2-methyl-5-trifluoromethoxy phenyl methanamine
PubChem CID 86277657
IUPAC Name [2-methyl-5-(trifluoromethoxy)phenyl]methanamine
SMILES CC1=C(CN)C=C(OC(F)(F)F)C=C1

Specifications

Quantity 1 g
UN Number UN3267
Sensitivity Air sensitive
Formula Weight 205.18
Percent Purity 96%
Chemical Name or Material 2-Methyl-5-(trifluoromethoxy)benzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only