missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7132820
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17161G 1 g
M17165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17161G Supplier TCI America Supplier No. M17161G
Only null left

Chemical Identifiers

CAS 1975-52-6
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007371
InChI Key DJRFJAVPROZZFL-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
PubChem CID 519683
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 1 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitrobenzoic Acid