accessibility menu, dialog, popup

UserName

CH3OONOO

CAS RN 1975-52-6

IUPAC Name: 2-methyl-5-nitrobenzoate
Synonyms: 2-methyl-5-nitrobenzoate
Molecular Weight (g/mol): 180.14
Molecular Formula: C8H6NO4
InChi Key: DJRFJAVPROZZFL-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1C([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)