missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic acid, 98+%

Catalog No. AAL1186206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1186206 5 g
AAL1186214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1186206 Supplier Thermo Scientific Chemicals Supplier No. L1186206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1975-52-6 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1975-52-6
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007371
InChI Key DJRFJAVPROZZFL-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
PubChem CID 519683
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 176°C to 181°C
Quantity 5 g
Beilstein 2451300
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-5-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-829-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only