missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7133074
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21785G 5 g
M217825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21785G Supplier TCI America Supplier No. M21785G
Only null left

Chemical Identifiers

CAS 121-02-8
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.64
MDL Number MFCD00051695
InChI Key WPGVQDHXOUAJBW-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzene-1-sulfonyl chloride, 5-nitro-o-toluenesulfonyl chloride, 2-methyl-5-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methyl-5-nitro, 4-nitrotoluen-2-sulfochlorid, o-toluenesulfonyl chloride, 5-nitro, 4-nitrotoluen-2-sulfonylchlorid, 4-nitrotoluene-2-sulphonyl chloride, 4-nitrotoluen-2-sulfochlorid czech, 2-methyl-5-nitrophenylsulfonyl chloride
PubChem CID 8457
IUPAC Name 2-methyl-5-nitrobenzene-1-sulfonyl chloride
SMILES CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 46°C
Color Yellow
Boiling Point 198°C
Quantity 5 g
UN Number 3261
Formula Weight 235.64
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methyl-5-nitrobenzenesulfonyl Chloride
RTECSNumber XT8000000
TSCA No