missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzenesulfonyl chloride, 97%

Catalog No. p-7022132
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1476006 5 g
AAA1476014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1476006 Supplier Thermo Scientific Chemicals Supplier No. A1476006
Only null left

CAS: 121-02-8 | C7H6ClNO4S | 235.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-02-8
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.64
MDL Number MFCD00051695
InChI Key WPGVQDHXOUAJBW-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzene-1-sulfonyl chloride, 5-nitro-o-toluenesulfonyl chloride, 2-methyl-5-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methyl-5-nitro, 4-nitrotoluen-2-sulfochlorid, o-toluenesulfonyl chloride, 5-nitro, 4-nitrotoluen-2-sulfonylchlorid, 4-nitrotoluene-2-sulphonyl chloride, 4-nitrotoluen-2-sulfochlorid czech, 2-methyl-5-nitrophenylsulfonyl chloride
PubChem CID 8457
IUPAC Name 2-methyl-5-nitrobenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 42°C to 46°C
Boiling Point 135°C to 137°C (0.7 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2697079
Sensitivity Moisture sensitive
Formula Weight 235.65
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-444-8
RTECSNumber XT8000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only