missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. M29575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29575G 5 g
M29571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29575G Supplier TCI America Supplier No. M29575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16634-91-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD03840506
InChI Key JLFWTLNLOSUFMU-UHFFFAOYSA-N
PubChem CID 12654145
IUPAC Name 2-methyl-5-nitrobenzaldehyde
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C=O

Specifications

Melting Point 55°C
Boiling Point 114°C
Quantity 5 g
Formula Weight 165.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitrobenzaldehyde