accessibility menu, dialog, popup

UserName

CH3NOOO

CAS RN 16634-91-6

IUPAC Name: 2-methyl-5-nitrobenzaldehyde
Synonyms: 2-methyl-5-nitrobenzaldehyde
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: JLFWTLNLOSUFMU-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results