Learn More
2-Methyl-3-nitrobenzotrifluoride, 97%
CAS: 6656-49-1 | C8H6F3NO2 | 205.14 g/mol
Supplier: Thermo Scientific Chemicals B2126714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methyl-3-nitrobenzotrifluoride | |
| 86°C (10 mmHg) | |
| C8H6F3NO2 | |
| MFCD00042322 | |
| UN2810 | |
| Light sensitive | |
| KQUQBPVYIURTNZ-UHFFFAOYSA-N | |
| 2-methyl-1-nitro-3-(trifluoromethyl)benzene | |
| 81171 | |
| 97% |
| 6656-49-1 | |
| >100°C (212°F) | |
| 1.478 | |
| 25 g | |
| 2457216 | |
| 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride | |
| CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O | |
| 205.14 | |
| 205.14 |
Chemical Identifiers
| 6656-49-1 | |
| 205.14 | |
| KQUQBPVYIURTNZ-UHFFFAOYSA-N | |
| 81171 | |
| CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O |
| C8H6F3NO2 | |
| MFCD00042322 | |
| 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride | |
| 2-methyl-1-nitro-3-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 229-688-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only