missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrobenzotrifluoride, 97%

Catalog No. p-7025653
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2126706 5 g
AAB2126714 25 g
2 options

Catalog No. AAB2126706

Supplier: Thermo Scientific Chemicals B2126706

Only null left

CAS: 6656-49-1 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6656-49-1
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD00042322
InChI Key KQUQBPVYIURTNZ-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
PubChem CID 81171
IUPAC Name 2-methyl-1-nitro-3-(trifluoromethyl)benzene
SMILES CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O

Specifications

Boiling Point 86°C (10 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.478
Quantity 5 g
UN Number UN2810
Beilstein 2457216
Sensitivity Light sensitive
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitrobenzotrifluoride
Hazard Category H301+H311-H315-H319
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 229-688-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only