missing translation for 'onlineSavingsMsg'
Learn More

2'-Methyl-3'-nitroacetanilide 98.0+%, TCI America™

Catalog No. p-7132791
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16515G 5 g
M165125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16515G Supplier TCI America Supplier No. M16515G
Only null left

Chemical Identifiers

CAS 56207-36-4
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD01047363
InChI Key NMYFXIITWKKOKY-UHFFFAOYSA-N
PubChem CID 151316
IUPAC Name N-(2-methyl-3-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Methyl-3′-nitroacetanilide