accessibility menu, dialog, popup

UserName

CAS RN 56207-36-4

CH3OHNH3CNOO

CAS RN 56207-36-4

IUPAC Name: N-(2-methyl-3-nitrophenyl)acetamide
Synonyms: N-(2-methyl-3-nitrophenyl)acetamide
Molecular Weight (g/mol): 194.19
Molecular Formula: C9H10N2O3
InChi Key: NMYFXIITWKKOKY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1C)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results