accessibility menu, dialog, popup

UserName

CH3H3COO(E)

CAS RN 3142-72-1

IUPAC Name: (2E)-2-methylpent-2-enoate
Synonyms: 2-methylpent-2-enoic acid
Molecular Weight (g/mol): 113.14
Molecular Formula: C6H9O2
InChi Key: JJYWRQLLQAKNAD-SNAWJCMRSA-M
SMILES: CC\C=C(/C)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)