missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-nitropropane 98.0+%, TCI America™
Supplier: TCI America M12415G
Specifications
| 2-Methyl-2-nitropropane | |
| White-Yellow | |
| MFCD00007393 | |
| 1325 | |
| AIMREYQYBFBEGQ-UHFFFAOYSA-N | |
| 2-methyl-2-nitropropane | |
| 11672 | |
| ≥98.0% (GC) |
| 594-70-7 | |
| C4H9NO2 | |
| 5 g | |
| propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044 | |
| CC(C)(C)[N+](=O)[O-] | |
| 103.121 | |
| 103.12 | |
| Crystals |
Chemical Identifiers
| 594-70-7 | |
| 103.121 | |
| AIMREYQYBFBEGQ-UHFFFAOYSA-N | |
| 11672 | |
| CC(C)(C)[N+](=O)[O-] |
| C4H9NO2 | |
| MFCD00007393 | |
| propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044 | |
| 2-methyl-2-nitropropane |
Safety and Handling
TSCA : Yes