missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-nitropropane 98.0+%, TCI America™

Catalog No. p-7132612
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12415G 5 g
M124125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12415G Supplier TCI America Supplier No. M12415G
Only null left

Chemical Identifiers

CAS 594-70-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00007393
InChI Key AIMREYQYBFBEGQ-UHFFFAOYSA-N
Synonym propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
PubChem CID 11672
IUPAC Name 2-methyl-2-nitropropane
SMILES CC(C)(C)[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 103.12
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Methyl-2-nitropropane