missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-adamantanol 99.0+%, TCI America™

$207.92 - $623.75

Chemical Identifiers

CAS 702-98-7
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00074811
InChI Key JKOZWMQUOWYZAB-UHFFFAOYSA-N
Synonym 2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane
PubChem CID 136545
IUPAC Name 2-methyladamantan-2-ol
SMILES CC1(O)C2CC3CC(C2)CC1C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13345G
View Documents
TCI America
M13345G
5 g
Each for $207.92
Only null left
 
M133425G
View Documents
TCI America
M133425G
25 g
Each for $623.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

702-98-7
166.26
JKOZWMQUOWYZAB-UHFFFAOYSA-N
136545
CC1(O)C2CC3CC(C2)CC1C3
C11H18O
MFCD00074811
2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane
2-methyladamantan-2-ol

Specifications

702-98-7
White
MFCD00074811
2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane
CC1(O)C2CC3CC(C2)CC1C3
166.26
166.26
Crystalline Powder
217°C
C11H18O
5 g
JKOZWMQUOWYZAB-UHFFFAOYSA-N
2-methyladamantan-2-ol
136545
≥99.0% (GC)
2-Methyl-2-adamantanol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1929

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'