missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-adamantanol 99.0+%, TCI America™
Supplier: TCI America M133425G
Specifications
| 2-Methyl-2-adamantanol | |
| 217°C | |
| C11H18O | |
| 25 g | |
| JKOZWMQUOWYZAB-UHFFFAOYSA-N | |
| 2-methyladamantan-2-ol | |
| 136545 | |
| ≥99.0% (GC) |
| 702-98-7 | |
| White | |
| MFCD00074811 | |
| 2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane | |
| CC1(O)C2CC3CC(C2)CC1C3 | |
| 166.26 | |
| 166.26 | |
| Crystalline Powder |
Chemical Identifiers
| 702-98-7 | |
| 166.26 | |
| JKOZWMQUOWYZAB-UHFFFAOYSA-N | |
| 136545 | |
| CC1(O)C2CC3CC(C2)CC1C3 |
| C11H18O | |
| MFCD00074811 | |
| 2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane | |
| 2-methyladamantan-2-ol |
Safety and Handling
EINECSNumber : (4)-1929
TSCA : No