accessibility menu, dialog, popup

UserName

CAS RN 702-98-7

CH3HO

CAS RN 702-98-7

IUPAC Name: 2-methyladamantan-2-ol
Synonyms: 2-methyladamantan-2-ol
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: JKOZWMQUOWYZAB-UHFFFAOYSA-N
SMILES: CC1(O)C2CC3CC(C2)CC1C3
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results