missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. p-4228819
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435561000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435561000 Supplier Thermo Scientific Chemicals Supplier No. 435561000
Only null left

CAS: 38614-36-7 | C4H7BrMg | 159.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38614-36-7 , 109-99-9
Molecular Formula C4H7BrMg
Molecular Weight (g/mol) 159.31
MDL Number MFCD00209600
InChI Key BKVIQODYSREJQH-UHFFFAOYSA-M
Synonym 2-methyl-1-propenylmagnesium bromide, 2-methyl-1-propenyl magnesium bromide, 2-methylpropenylmagnesium bromide, 2-methyl-propenylmagnesium bromide, 2-methylpropenyl magnesium bromide, bkviqodysrejqh-uhfffaoysa-m, 2-methyl propenyl magnesium bromide, 2-methylprop-1-enylmagnesium bromide, 2-methylprop-1-enyl magnesium bromide, grignard reagent
PubChem CID 4456485
IUPAC Name magnesium(2+) 2-methylprop-1-en-1-ide bromide
SMILES CC(C)=C[Mg]Br

Specifications

CAS Min % 7.53, 90.8
CAS Max % 9.2, 92.47
Color Orange to Yellow
Density 0.9520g/mL
Flash Point −15°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.952
Solubility Information Solubility in water: reacts
Formula Weight 159.31
Physical Form Solution
Chemical Name or Material 2-Methyl-1-propenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Suspected of causing cancer if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flamma
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only