CAS RN 38614-36-7
CAS RN 38614-36-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
CAS: 38614-36-7 Molecular Formula: C4H7BrMg Molecular Weight (g/mol): 159.31 MDL Number: MFCD00209600 InChI Key: BKVIQODYSREJQH-UHFFFAOYSA-M Synonym: 2-methyl-1-propenylmagnesium bromide,2-methyl-1-propenyl magnesium bromide,2-methylpropenylmagnesium bromide,2-methyl-propenylmagnesium bromide,2-methylpropenyl magnesium bromide,bkviqodysrejqh-uhfffaoysa-m,2-methyl propenyl magnesium bromide,2-methylprop-1-enylmagnesium bromide,2-methylprop-1-enyl magnesium bromide,grignard reagent PubChem CID: 4456485 IUPAC Name: magnesium(2+) 2-methylprop-1-en-1-ide bromide SMILES: CC(C)=C[Mg]Br