Learn More
2-Methyl-1-propanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™
2-Methyl-1-propanol can be obtained via hydroformylation of alkenes with fuel gas mixture in particular, carbon monoxide being the main reactant.
Supplier: Merck Emd Millipore 8205910MLF
Specifications
| 78-83-1 | |
| 2.55 (vs. air) | |
| Neat | |
| MFCD00004740 | |
| 108°C | |
| 10 mL | |
| ≥99.8% (GC) |
| 8.8 mmHg (0°C) | |
| 0.803 g/mL (at 25°C) | |
| (CH3)2 CHCH2OH | |
| -108°C | |
| Isobutanol; Isobutyl alcohol | |
| 74.12 | |
| Analytical Standard |