missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-phenyl-2-butanol 98.0+%, TCI America™
Supplier: TCI America M201025G
Specifications
| 2-Methyl-1-phenyl-2-butanol | |
| Yellow | |
| C11H16O | |
| 25 g | |
| CCC(C)(O)CC1=CC=CC=C1 | |
| 164.25 | |
| 164.25 | |
| Liquid |
| 772-46-3 | |
| 61°C | |
| MFCD01708473 | |
| FTZBYXCNXOPJEL-UHFFFAOYNA-N | |
| 2-methyl-1-phenylbutan-2-ol | |
| 101135 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 772-46-3 | |
| 164.25 | |
| FTZBYXCNXOPJEL-UHFFFAOYNA-N | |
| 2-methyl-1-phenylbutan-2-ol |
| C11H16O | |
| MFCD01708473 | |
| 101135 | |
| CCC(C)(O)CC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1034
RTECSNumber : SG8280000
TSCA : No