accessibility menu, dialog, popup

UserName

CH3H3COH

CAS RN 772-46-3

IUPAC Name: 2-methyl-1-phenylbutan-2-ol
Synonyms: 2-methyl-1-phenylbutan-2-ol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: FTZBYXCNXOPJEL-UHFFFAOYNA-N
SMILES: CCC(C)(O)CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results