missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-2-butanol 98.0+%, TCI America™

Catalog No. M20105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20105G 5 g
M201025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20105G Supplier TCI America Supplier No. M20105G

Chemical Identifiers

CAS 772-46-3
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD01708473
InChI Key FTZBYXCNXOPJEL-UHFFFAOYNA-N
PubChem CID 101135
IUPAC Name 2-methyl-1-phenylbutan-2-ol
SMILES CCC(C)(O)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 61°C
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-phenyl-2-butanol
EINECSNumber (3)-1034
RTECSNumber SG8280000
TSCA No